Assay Details
Gene Symbol: Pmepa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107807.2 | NP_001101277.1 | 2 - 3 | 172 | 67 | |
XM_006235689.1 | XP_006235751.1 | 3 - 4 | 337 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
negative regulation of SMAD protein complex assembly
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of pathway-restricted SMAD protein phosphorylation
molecular_function
WW domain binding
R-SMAD binding
biological_process
negative regulation of SMAD protein complex assembly
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of pathway-restricted SMAD protein phosphorylation
molecular_function
WW domain binding
R-SMAD binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002899 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158211 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002899 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002904 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002899 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002904 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002899 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002904 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P426788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92517 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10852146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852145 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10852144 | - | - |
Rat Genome 230 2.0 Array | 1392534_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389809_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392534_at | - | - |
Rat Genome 230 2.0 Array | 1389809_at | - | - |
Affymetrix HT Rat Focus Array | 1392534_at | - | - |
Affymetrix HT Rat Focus Array | 1392534_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300037705.rn.1 | TC0300004948.rn.1 | RTA TC |
Rat Expression Set 230 | 1392534_at | - | - |
Rat Expression Set 230 | 1389809_at | - | - |
Rat Expression Set 230 | 1389809_PM_at | - | - |
Rat Expression Set 230 | 1392534_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10852144 | - | - |
Rat Genome U34 Set | rc_AI639058_s_at | - | - |
Rat Genome U34 Set | rc_AI058507_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364908 | - | - |
