Assay Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255011.2 | XP_006255073.1 | 21 - 22 | 3736 | 60 | |
XM_006255013.2 | XP_006255075.1 | 21 - 22 | 3736 | 60 | ||
XM_006255016.2 | XP_006255078.1 | 21 - 22 | 3736 | 60 | ||
XM_008772218.1 | XP_008770440.1 | 21 - 22 | 3870 | 60 | ||
XM_008772219.1 | XP_008770441.1 | 21 - 22 | 3736 | 60 | ||
XM_008772220.1 | XP_008770442.1 | 21 - 22 | 3736 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
