Assay Details
| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191814.1 | NP_001178743.1 | 5 - 6 | 1074 | 72 | |
| XM_006245658.2 | XP_006245720.1 | 6 - 7 | 1732 | 72 | ||
| XM_006245661.2 | XP_006245723.1 | 5 - 6 | 1670 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl4
Genomic Map