Assay Details
| Gene Symbol: Cenpw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246319.1 | NP_001233248.1 | 2 - 3 | 349 | 94 | |
| GenBank mRNA | BC129114.1 | - | 2 - 3 | 349 | 7112235 | 94 |
| FQ220188.1 | - | 1 - 2 | 78 | 94 | ||
| FQ227404.1 | - | 1 - 2 | 78 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cenpw
Genomic Map