Assay Details
Gene Symbol: RT1-CE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002155.1 | 1 | 618 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RT1 class I, locus1 |
RT1-CE1 | RT1-C/E1 | Sense | Chr. 20 - 4638235 - 4792390 on NCBI Build Rnor_6.0 | |
Downstream |
RT1 class I, locus CE7 |
RT1-CE7 | RT1-C/E7 | 18.0kb | Sense | Chr. 20 - 4695765 - 4698712 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
