Assay Details
| Gene Symbol: LOC686120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026689.1 | 2 - 3 | 155 | 81 | ||
| GenBank mRNA | CB705680.1 | - | 2 - 3 | 155 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | syntaxin 1A (brain) | Stx1a | -11.5kb | Anti-sense | Chr. 12 - 24682046 - 24710047 on NCBI Build Rnor_6.0 | |
| Downstream | abhydrolase domain containing 11 | Abhd11 |
Abhd11l1, Wbscr21 |
392.0 | Anti-sense | Chr. 12 - 24722277 - 24724997 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC686120
Genomic Map