Assay Details
| Gene Symbol: Tssk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073162.1 | 2 - 3 | 591 | 97 | ||
| GenBank mRNA | BC079202.1 | - | 2 - 3 | 591 | 7131733 | 97 |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CK598791.1 | - | 590 | Chr. - - [-] on Build | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane 9 superfamily member 1 | Tm9sf1 | -7.1kb | Anti-sense | Chr. 15 - 34314645 - 34322390 on NCBI Build Rnor_6.0 | |
| Downstream | magnesium-dependent phosphatase 1 | Mdp1 |
Mdp-1, RGD1311147 |
5.6kb | Anti-sense | Chr. 15 - 34336058 - 34340084 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

Tssk4
Genomic Map