Assay Details
Gene Symbol: Skp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007608.2 | NP_001007609.1 | 2 - 3 | 179 | 67 | |
XM_006246244.2 | XP_006246306.1 | 2 - 3 | 478 | 67 | ||
GenBank mRNA | BC058152.1 | - | 2 - 3 | 182 | 6890712 | 67 |
FQ210698.1 | - | 1 - 2 | 77 | 67 | ||
FQ212502.1 | - | 1 - 2 | 77 | 67 | ||
FQ212528.1 | - | 1 - 2 | 77 | 67 | ||
FQ212745.1 | - | 1 - 2 | 77 | 67 | ||
FQ212916.1 | - | 1 - 2 | 77 | 67 | ||
FQ213991.1 | - | 1 - 2 | 77 | 67 | ||
FQ214002.1 | - | 1 - 2 | 77 | 67 | ||
FQ214005.1 | - | 1 - 2 | 77 | 67 | ||
FQ214369.1 | - | 1 - 2 | 77 | 67 | ||
FQ214470.1 | - | 1 - 2 | 77 | 67 | ||
FQ214982.1 | - | 1 - 2 | 77 | 67 | ||
FQ215334.1 | - | 1 - 2 | 77 | 67 | ||
FQ215452.1 | - | 1 - 2 | 77 | 67 | ||
FQ219401.1 | - | 1 - 2 | 77 | 67 | ||
FQ224935.1 | - | 1 - 2 | 77 | 67 | ||
FQ229788.1 | - | 77 | 67 | |||
FQ230985.1 | - | 1 - 2 | 77 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
