Assay Details
| Gene Symbol: Faim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014843.2 | NP_001014843.1 | 7 - 8 | 1196 | 107 | |
| GenBank mRNA | BC088197.1 | - | 7 - 8 | 1196 | 7375085 | 107 |
| FQ232848.1 | - | 4 - 5 | 640 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faim3
Genomic Map