Assay Details
Gene Symbol: Chst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037775.2 | NP_001032864.1 | 1 | 1021 | 56 | |
XM_006248935.1 | XP_006248997.1 | 2 | 1115 | 56 | ||
XM_006248936.2 | XP_006248998.1 | 2 | 2164 | 56 | ||
XM_006248937.2 | XP_006248999.1 | 2 | 2902 | 56 | ||
XM_008768971.1 | XP_008767193.1 | 3 | 2046 | 56 | ||
GenBank mRNA | BC100096.1 | - | 1 | 1025 | 7375118 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
