Assay Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080907.2 | NP_001074376.1 | 3 - 4 | 342 | 74 | |
XM_006232254.2 | XP_006232316.1 | 4 - 5 | 412 | 74 | ||
XM_008760942.1 | XP_008759164.1 | 3 - 4 | 1891 | 74 | ||
XM_008760943.1 | XP_008759165.1 | 4 - 5 | 1163 | 74 | ||
GenBank mRNA | BC087704.1 | - | 3 - 4 | 343 | 7313723 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
