Assay Details
| Gene Symbol: Kcnmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104560.2 | NP_001098030.1 | 1 - 2 | 321 | 109 | |
| GenBank mRNA | AB297662.1 | - | 1 - 2 | 322 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnmb3
Genomic Map