Assay Details
| Gene Symbol: Sftpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270645.1 | NP_001257574.1 | 3 - 4 | 353 | 78 | |
| NM_001270647.1 | NP_001257576.1 | 3 - 4 | 564 | 78 | ||
| NM_017329.2 | NP_059025.2 | 3 - 4 | 350 | 78 | ||
| GenBank mRNA | BC085353.1 | - | 3 - 4 | 353 | 7121530 | 78 |
| CK364490.1 | - | 3 - 4 | 576 | 78 | ||
| M15754.1 | - | 3 - 4 | 338 | 78 | ||
More Information
Set Membership: |
|

Sftpa1
Genomic Map