Assay Details
| Gene Symbol: Sftpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270645.1 | NP_001257574.1 | 4 - 5 | 436 | 92 | |
| NM_001270647.1 | NP_001257576.1 | 4 - 5 | 647 | 92 | ||
| NM_017329.2 | NP_059025.2 | 4 - 5 | 433 | 92 | ||
| GenBank mRNA | BC085353.1 | - | 4 - 5 | 436 | 7121530 | 92 |
| CK364490.1 | - | 4 - 5 | 659 | 92 | ||
| M15754.1 | - | 4 - 5 | 421 | 92 | ||
| M33201.1 | - | 4 - 5 | 430 | 92 | ||
| X13176.1 | - | 4 - 5 | 421 | 92 | ||
| X13177.1 | - | 4 - 5 | 430 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sftpa1
Genomic Map