Assay Details
| Gene Symbol: LOC100910996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271382.1 | NP_001258311.1 | 5 - 6 | 224 | 64 | |
| XM_008765212.1 | XP_008763434.1 | 5 - 6 | 376 | 64 | ||
| GenBank mRNA | FQ222373.1 | - | 1 - 2 | 31 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910996
Genomic Map