Assay Details
| Gene Symbol: Zfp496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271396.1 | NP_001258325.1 | 6 - 7 | 1041 | 82 | |
| XM_006246450.1 | XP_006246512.1 | 6 - 7 | 1187 | 82 | ||
| XM_006246451.1 | XP_006246513.1 | 6 - 7 | 1029 | 82 | ||
| XM_006246452.1 | XP_006246514.1 | 6 - 7 | 1043 | 82 | ||
| XM_008767781.1 | XP_008766003.1 | 3 - 4 | 339 | 82 | ||
| GenBank mRNA | FQ223523.1 | - | 6 - 7 | 1043 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp496
Genomic Map