Assay Details
| Gene Symbol: Zfp496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271396.1 | NP_001258325.1 | 7 - 8 | 1168 | 92 | |
| XM_006246450.1 | XP_006246512.1 | 7 - 8 | 1314 | 92 | ||
| XM_006246451.1 | XP_006246513.1 | 7 - 8 | 1156 | 92 | ||
| XM_006246452.1 | XP_006246514.1 | 7 - 8 | 1170 | 92 | ||
| XM_008767781.1 | XP_008766003.1 | 4 - 5 | 466 | 92 | ||
| GenBank mRNA | FQ223523.1 | - | 7 - 8 | 1170 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp496
Genomic Map