Assay Details
| Gene Symbol: Fabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012556.2 | NP_036688.1 | 1 - 2 | 108 | 72 | |
| GenBank mRNA | BC086947.1 | - | 1 - 2 | 108 | 7325407 | 72 |
| FQ209859.1 | - | 1 - 2 | 110 | 72 | ||
| FQ209983.1 | - | 1 - 2 | 117 | 72 | ||
| FQ210522.1 | - | 1 - 2 | 117 | 72 | ||
| FQ211310.1 | - | 1 - 2 | 117 | 72 | ||
| FQ211342.1 | - | 1 - 2 | 78 | 72 | ||
| M17899.1 | - | 1 - 2 | 76 | 72 | ||
| M35991.1 | - | 1 - 2 | 109 | 72 | ||
| V01235.1 | - | 1 - 2 | 109 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp1
Genomic Map