Assay Details
| Gene Symbol: Bet1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019368.3 | NP_062241.2 | 2 - 3 | 161 | 63 | |
| XM_006230574.2 | XP_006230636.1 | 3 - 4 | 373 | 63 | ||
| GenBank mRNA | AF003998.1 | - | 2 - 3 | 200 | 63 | |
| BC059138.1 | - | 2 - 3 | 165 | 6890305 | 63 | |
| FQ228853.1 | - | 3 - 4 | 323 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bet1l
Genomic Map