Assay Details
| Gene Symbol: Ugt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031533.5 | NP_113721.4 | 1 - 2 | 755 | 75 | |
| GenBank mRNA | BC089792.1 | - | 1 - 2 | 738 | 7370057 | 75 |
| DQ351215.1 | - | 1 - 2 | 1096 | 75 | ||
| EX489691.1 | - | 1 - 2 | 322 | 75 | ||
| X03478.1 | - | 1 - 2 | 644 | 75 | ||
| Gene Symbol: Ugt2b17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769997.1 | XP_008768219.1 | 1 - 2 | 1095 | 75 | |
| XM_008769999.1 | XP_008768221.1 | 1 - 2 | 749 | 75 | ||
| XM_008770000.1 | XP_008768222.1 | 1 - 2 | 749 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ugt2b
Genomic Map