Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052798.2 | NP_434685.1 | 1 - 2 | 339 | 66 | |
| XM_006246227.2 | XP_006246289.1 | 2 - 3 | 1486 | 66 | ||
| XM_008767660.1 | XP_008765882.1 | 2 - 3 | 1486 | 66 | ||
| XM_008767661.1 | XP_008765883.1 | 2 - 3 | 1486 | 66 | ||
| GenBank mRNA | BC078777.1 | - | 2 - 3 | 176 | 7127868 | 66 |
| M96548.1 | - | 1 - 2 | 341 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp354a
Genomic Map