Assay Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130400.2 | NP_569084.1 | 1 - 2 | 171 | 70 | |
| GenBank mRNA | AF318150.1 | - | 1 - 2 | 172 | 70 | |
| BC158583.1 | - | 98 | 9022774 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dhfr
Genomic Map