Assay Details
| Gene Symbol: Mir6323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106695.1 | 1 | 51 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kelch-like family member 6 | Klhl6 | -41.7kb | Sense | Chr. 11 - 84745958 - 84785315 on NCBI Build Rnor_6.0 | |
| Downstream | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 | B3gnt5 | 91.9kb | Anti-sense | Chr. 11 - 84919038 - 84931160 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6323
Genomic Map