Assay Details
| Gene Symbol: Mir6333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106707.1 | 1 | 52 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NIPA-like domain containing 3 | Nipal3 |
Npal3, RGD1563439 |
Anti-sense | Chr. 5 - 153808181 - 153843951 on NCBI Build Rnor_6.0 | |
| Downstream | sperm-tail PG-rich repeat containing 1 | Stpg1 | Mapo2 | 28.0 | Sense | Chr. 5 - 153844039 - 153888510 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6333
Genomic Map