Assay Details
| Gene Symbol: LOC100910558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110678.1 | 3 - 4 | 605 | 63 | ||
| GenBank mRNA | FM052678.1 | - | 3 - 4 | 356 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor I/B | Nfib | -37.1kb | Anti-sense | Chr. 5 - 100433268 - 100647963 on NCBI Build Rnor_6.0 | |
| Downstream |
zinc finger, DHHC-type containing 21 |
Zdhhc21 | 236.4kb | Anti-sense | Chr. 5 - 100924621 - 100983380 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910558
Genomic Map