Assay Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277228.1 | NP_001264157.1 | 9 - 10 | 1762 | 85 | |
| GenBank mRNA | BC085943.1 | - | 8 - 9 | 1882 | 85 | |
| BC168987.1 | - | 9 - 10 | 1724 | 9030170 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kdelc1
Genomic Map