Assay Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC101929.1 | - | 4 - 5 | 641 | 7939237 | 96 |
| CK222497.1 | - | 3 - 4 | 96 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cox15
Genomic Map