Assay Details
| Gene Symbol: Snrpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271237.1 | NP_001258166.1 | 3 - 4 | 160 | 70 | |
| GenBank mRNA | FQ211122.1 | - | 3 - 4 | 160 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snrpe
Genomic Map