Assay Details
| Gene Symbol: Timm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276429.1 | NP_001263358.1 | 4 - 5 | 308 | 116 | |
| NM_001276430.1 | NP_001263359.1 | 3 - 4 | 293 | 116 | ||
| NM_133604.1 | NP_598288.1 | 4 - 5 | 379 | 116 | ||
| GenBank mRNA | FM052814.1 | - | 4 - 5 | 217 | 116 | |
| FM056491.1 | - | 3 - 4 | 256 | 116 | ||
| FQ228524.1 | - | 4 - 5 | 379 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm9
Genomic Map