Assay Details
| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134704.1 | NP_001128176.1 | 354 | 63 | ||
| GenBank mRNA | BC166795.1 | - | 354 | 7937873 | 63 | |
More Information
Set Membership: |
|

Ormdl1
Genomic Map