Assay Details
Gene Symbol: Nfe2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_231763.8 | XP_231763.6 | 954 | 96 | ||
GenBank mRNA | DY310098.1 | - | 599 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
