Assay Details
| Gene Symbol: Hist1h2ah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253977.2 | XP_006254039.1 | 1 | 445 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ah| Gene Symbol: Hist1h2ah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253977.2 | XP_006254039.1 | 1 | 445 | 71 | |
Set Membership: |
|