Assay Details
| Gene Symbol: LOC100364523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771462.1 | XP_008769684.1 | 1 - 2 | 48 | 70 | |
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC107913.1 | - | 48 | 7460613 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107913.1, XM_008771462.1 transcripts.
Set Membership: |
|

LOC100364523
Genomic Map