Assay Details
| Gene Symbol: LOC100362224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FQ228280.1 | - | 74 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | karyopherin alpha 4 (importin alpha 3) | Kpna4 | importin | -19.5kb | Anti-sense | Chr. 2 - 165684345 - 165739874 on NCBI Build Rnor_6.0 |
| Downstream | similar to Mss4 protein | RGD1563962 | 37.7kb | Sense | Chr. 2 - 165805230 - 165816503 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC100362224
Genomic Map