Assay Details
Gene Symbol: LOC103692944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC088332.1 | - | 1 - 2 | 201 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | somatostatin receptor 3 | Sstr3 |
SSTR, Smstr28 |
-10.9kb | Sense | Chr. 7 - 119760881 - 119768082 on NCBI Build Rnor_6.0 |
Downstream |
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2) |
Rac2 | 4.3kb | Sense | Chr. 7 - 119783872 - 119797111 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
