Assay Details
| Gene Symbol: Cct7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106603.1 | NP_001100073.1 | 12 | 2245 | 106 | |
| GenBank mRNA | FQ214319.1 | - | 12 | 1615 | 106 | |
More Information
Set Membership: |
|

Cct7
Genomic Map