Assay Details
| Gene Symbol: Cox8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134345.1 | NP_599172.1 | 2 | 405 | 71 | |
| GenBank mRNA | BC126066.1 | - | 2 | 372 | 7385046 | 71 |
| L48209.1 | - | 2 | 323 | 71 | ||
| M28255.1 | - | 1 | 244 | 71 | ||
| X06146.1 | - | 250 | 71 | |||
More Information
Set Membership: |
|

Cox8a
Genomic Map