Assay Details
| Gene Symbol: LOC102547035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254826.2 | XP_006254888.1 | 1 - 2 | 145 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102547035| Gene Symbol: LOC102547035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254826.2 | XP_006254888.1 | 1 - 2 | 145 | 75 | |
Set Membership: |
|