Assay Details
Gene Symbol: Zfp238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250328.2 | XP_006250390.1 | 3 | 4318 | 82 | |
XM_006250330.2 | XP_006250392.1 | 2 | 4234 | 82 | ||
XM_006250331.2 | XP_006250393.1 | 2 | 3713 | 82 | ||
XM_008769796.1 | XP_008768018.1 | 2 | 4965 | 82 | ||
XM_008769797.1 | XP_008768019.1 | 3 | 4097 | 82 | ||
XM_008769798.1 | XP_008768020.1 | 3 | 4317 | 82 | ||
XM_008769799.1 | XP_008768021.1 | 2 | 6236 | 82 | ||
XM_008769800.1 | XP_008768022.1 | 2 | 3268 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
