Assay Details
| Gene Symbol: RGD1560801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761545.1 | XP_008759767.1 | 13 - 14 | 2249 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560801| Gene Symbol: RGD1560801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761545.1 | XP_008759767.1 | 13 - 14 | 2249 | 135 | |
Set Membership: |
|