Assay Details
| Gene Symbol: LOC100910256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750492.2 | XP_003750540.2 | 820 | 97 | ||
| Gene Symbol: LOC100910010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749260.3 | XP_003749308.3 | 1 | 820 | 97 | |
| Gene Symbol: LOC100910041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749182.3 | XP_003749230.3 | 1 | 820 | 97 | |
| Gene Symbol: LOC100910335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750494.2 | XP_003750542.2 | 1 | 820 | 97 | |
| Gene Symbol: LOC100910445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750496.3 | XP_003750544.3 | 1 | 820 | 97 | |
| Gene Symbol: Olr1275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001000800.1 | NP_001000800.1 | 820 | 97 | ||
More Information
Set Membership: |
|

LOC100910256
Genomic Map