Assay Details
| Gene Symbol: RGD1564162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256332.2 | XP_006256394.1 | 1 - 2 | 1256 | 74 | |
| XM_006256333.2 | XP_006256395.1 | 1 - 2 | 1256 | 74 | ||
| XM_006256335.2 | XP_006256397.1 | 1 - 2 | 1256 | 74 | ||
| XM_006256336.2 | XP_006256398.1 | 1 - 2 | 1256 | 74 | ||
| XM_008772934.1 | XP_008771156.1 | 1 - 2 | 1256 | 74 | ||
| XM_008772935.1 | XP_008771157.1 | 1 - 2 | 1256 | 74 | ||
| GenBank mRNA | BC160832.1 | - | 1 - 2 | 520 | 74 | |
| Gene Symbol: LOC103694873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773822.1 | XP_008772044.1 | 1 - 2 | 1256 | 74 | |
| XM_008773823.1 | XP_008772045.1 | 1 - 2 | 1256 | 74 | ||
| XM_008773824.1 | XP_008772046.1 | 1 - 2 | 1256 | 74 | ||
| XM_008773825.1 | XP_008772047.1 | 1 - 2 | 1256 | 74 | ||
| XM_008773826.1 | XP_008772048.1 | 1 - 2 | 1256 | 74 | ||
| XM_008773827.1 | XP_008772049.1 | 1 - 2 | 1256 | 74 | ||
More Information
Set Membership: |
|

RGD1564162
Genomic Map