Assay Details
| Gene Symbol: Tceb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 525 | 67 | |
| NM_001270562.1 | NP_001257491.1 | 4 | 423 | 67 | ||
| NM_001270563.1 | NP_001257492.1 | 4 | 383 | 67 | ||
| NM_022593.4 | NP_072115.1 | 4 | 419 | 67 | ||
| XM_006237724.2 | XP_006237786.1 | 4 | 420 | 67 | ||
| GenBank mRNA | BC060566.1 | - | 4 | 394 | 6918038 | 67 |
| CK355540.1 | - | 4 | 419 | 67 | ||
| FM093546.1 | - | 4 | 385 | 67 | ||
| FM101371.1 | - | 4 | 373 | 67 | ||
| FM110992.1 | - | 4 | 389 | 67 | ||
| FQ213660.1 | - | 4 | 433 | 67 | ||
| FQ214805.1 | - | 4 | 525 | 67 | ||
| L29259.1 | - | 3 | 336 | 67 | ||
More Information
Set Membership: |
|

Tceb1
Genomic Map