Assay Details
| Gene Symbol: LOC100909869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256328.2 | XP_006256390.1 | 3 | 1006 | 115 | |
| Gene Symbol: LOC100909461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773851.1 | XP_008772073.1 | 1 | 2161 | 115 | |
| Gene Symbol: LOC103690173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773852.1 | XP_008772074.1 | 3 | 1006 | 115 | |
| Gene Symbol: Zfp280b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106384.1 | NP_001099854.1 | 2 | 917 | 115 | |
| XM_008772876.1 | XP_008771098.1 | 3 | 1006 | 115 | ||
More Information
Set Membership: |
|

LOC100909869
Genomic Map