Assay Details
| Gene Symbol: LOC102554393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233879.2 | XP_006233941.2 | 17 - 18 | 2183 | 62 | |
| Gene Symbol: LOC102554283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233729.2 | XP_006233791.1 | 2 - 3 | 951 | 62 | |
More Information
Set Membership: |
|

LOC102554393
Genomic Map