Assay Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235009.2 | XP_006235071.1 | 1 | 657 | 60 | |
| XM_006235011.2 | XP_006235073.1 | 1 | 657 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235009.2, XM_006235011.2 transcripts.
Set Membership: |
|

Lzts3
Genomic Map