Assay Details
| Gene Symbol: LOC102546678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228173.2 | XP_006228235.1 | 2 - 3 | 137 | 59 | |
| XM_006228174.1 | XP_006228236.1 | 2 - 3 | 335 | 59 | ||
| XM_006228175.2 | XP_006228237.1 | 2 - 3 | 117 | 59 | ||
| XM_008758995.1 | XP_008757217.1 | 2 - 3 | 235 | 59 | ||
| XM_008758996.1 | XP_008757218.1 | 2 - 3 | 234 | 59 | ||
| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253297.1 | XP_006253359.1 | 2 - 3 | 335 | 59 | |
| XM_006253298.2 | XP_006253360.1 | 2 - 3 | 117 | 59 | ||
| XM_008771339.1 | XP_008769561.1 | 2 - 3 | 235 | 59 | ||
| XM_008771340.1 | XP_008769562.1 | 2 - 3 | 234 | 59 | ||
| XM_219845.8 | XP_219845.5 | 2 - 3 | 137 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228173.2, XM_006228174.1, XM_006228175.2, XM_006253297.1, XM_006253298.2, XM_008758995.1, XM_008758996.1, XM_008771339.1, XM_008771340.1, XM_219845.8 transcripts.
Set Membership: |
|

LOC102546678
Genomic Map