Assay Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100565.1 | NP_001094035.1 | 7 | 2010 | 98 | |
| GenBank mRNA | BC092604.1 | - | 1 | 840 | 98 | |
| BC166745.1 | - | 7 | 1843 | 7939059 | 98 | |
More Information
Set Membership: |
|

Peli1
Genomic Map