Assay Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729406.4 | XP_002729452.2 | 1 | 391 | 93 | |
| XM_006237213.2 | XP_006237275.1 | 1 | 392 | 93 | ||
| XM_006237215.2 | XP_006237277.1 | 1 | 391 | 93 | ||
| XM_008763270.1 | XP_008761492.1 | 1 | 391 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002729406.4, XM_006237213.2, XM_006237215.2, XM_008763270.1 transcripts.
Set Membership: |
|

Zfp248
Genomic Map