Assay Details
Gene Symbol: Tas2r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237482.1 | XP_006237544.1 | 1 | 369 | 76 |
Gene Symbol: Tas2r13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023998.1 | NP_076488.1 | 1 | 309 | 76 | |
GenBank mRNA | AF227145.1 | - | 1 | 309 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
